Assay Details
| Gene Symbol: Fbxo44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK170958.1 | - | 2 - 3 | 322 | 72 | |
| AK191511.1 | - | 1 - 2 | 235 | 72 | ||
| AK218805.1 | - | 1 - 2 | 168 | 72 | ||
More Information
Set Membership: |
|

Fbxo44
Genomic Map