Assay Details
| Gene Symbol: Fbxo44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161851.2 | NP_001155323.1 | 5 - 6 | 724 | 64 | |
| NM_001161852.2 | NP_001155324.1 | 4 - 5 | 876 | 64 | ||
| NM_173401.5 | NP_775577.2 | 5 - 6 | 1001 | 64 | ||
| GenBank mRNA | CA322196.1 | - | 2 - 3 | 180 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23392753 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23392750 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23392754 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23392749 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23392756 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23392752 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054350 | AK218373.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054350 | BY658029.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054350 | NM_173401.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054350 | AK077452.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054350 | AK170958.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2085333 | AK191511.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2085333 | AK218805.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2085333 | BC028884.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2085333 | NM_173401.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2085333 | AK019407.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 549195 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 859477 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 452270 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 801621 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 130117 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P156523 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054350 | AK218373.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054350 | BY658029.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054350 | NM_173401.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054350 | AK077452.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054350 | AK170958.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2085333 | AK191511.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2085333 | AK218805.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2085333 | BC028884.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2085333 | NM_173401.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2085333 | AK019407.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392747 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392741 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392734 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392738 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392732 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392740 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392725 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392737 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392730 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392752 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392742 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392753 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392736 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392710 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392716 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392711 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392721 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392727 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392749 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392713 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392743 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392708 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392745 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392748 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392750 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392729 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392712 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392754 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392719 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392723 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392751 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392714 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P156523 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01013343 | - | - |
| Whole Mouse Genome G4122A | A_52_P651498 | BC028884.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P651498 | NM_173401.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P651498 | AK019407.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P651498 | AK077452.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P651498 | NM_001161851.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P505943 | AK077452.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111355 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P156523 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03975322 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03975335 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03445824 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03975340 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03975331 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23392754 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23392753 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23392756 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23392749 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23392750 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23392752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392657 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392710 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392747 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392732 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392713 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392745 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03445815 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392727 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03975336 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03975342 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392736 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392725 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03445823 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392721 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392723 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392734 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392729 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392716 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03975340 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392708 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04016736 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392712 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392742 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392749 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04031386 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392743 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392748 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392752 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10518486 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10518487 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10518485 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10518491 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10518488 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10518490 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10518489 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430541_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434455_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10518484 | - | - |
| Mouse Genome 430 2.0 Array | 1430541_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1434455_at | - | - |
| Mouse Genome 430 2.0 Array | 1430541_at | - | - |
| Mouse Genome 430 2.0 Array | 1434455_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 230903_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 230903_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400033208.mm.1 | NM_001161851.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400033208.mm.1 | NM_001161852.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400033208.mm.1 | NM_173401.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400033208.mm.1 | NR_104374.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400033208.mm.1 | NR_104375.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400033208.mm.1 | TC0400004016.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400033209.mm.1 | NM_001161851.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400033209.mm.1 | NM_001161852.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400033209.mm.1 | NM_173401.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400033209.mm.1 | NR_104374.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400033209.mm.1 | NR_104375.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400033209.mm.1 | TC0400004016.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400033207.mm.1 | NM_001161851.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400033207.mm.1 | NM_001161852.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400033207.mm.1 | NM_173401.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400033207.mm.1 | NR_104374.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400033207.mm.1 | NR_104375.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400033207.mm.1 | TC0400004016.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400033210.mm.1 | NM_001161851.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400033210.mm.1 | NM_001161852.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400033210.mm.1 | NM_173401.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400033210.mm.1 | NR_104374.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400033210.mm.1 | NR_104375.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400033210.mm.1 | TC0400004016.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1434455_PM_at | - | - |
| Mouse Expression Set 430 | 1434455_at | - | - |
| Mouse Expression Set 430 | 1430541_PM_at | - | - |
| Mouse Expression Set 430 | 1430541_at | - | - |
| Murine Genome U74v2 Set | 165672_PM_at | - | - |
| Murine Genome U74v2 Set | 165672_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430541_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430541_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 104560494 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 104560494 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4760372 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004760372 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1229019 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1230435 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1229019 | - | - |
| mouse-6 v1.1 (Illumina) | scl23522.8_437-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4760372 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 104560494 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl23522.8_437-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4760372 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104560494 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1229019 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1229019 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2550725 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl23522.8_437-S | - | - |

Fbxo44
Genomic Map