Assay Details
| Gene Symbol: Fbxo44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161852.2 | NP_001155324.1 | 3 - 4 | 735 | 73 | |
| GenBank mRNA | BM932876.1 | - | 2 - 3 | 384 | 73 | |
| BQ042735.1 | - | 3 - 4 | 518 | 73 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Fbxo44
Genomic Map