Assay Details
| Gene Symbol: Samd11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110516.1 | NP_001103986.1 | 9 - 10 | 91 | ||
| GenBank mRNA | AK149268.1 | - | 9 - 10 | 91 | ||
| AK161035.1 | - | 8 - 9 | 91 | |||
| AY458844.1 | - | 8 - 9 | 91 | |||
| BC026511.1 | - | 4 - 5 | 91 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Samd11
Genomic Map