Assay Details
| Gene Symbol: Noc2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021303.2 | NP_067278.2 | 16 - 17 | 1963 | 71 | |
| GenBank mRNA | AF155546.1 | - | 4 - 5 | 473 | 71 | |
| AK076287.1 | - | 15 - 16 | 2005 | 71 | ||
| AK078560.1 | - | 16 - 17 | 1941 | 71 | ||
| AK133834.1 | - | 16 - 17 | 1963 | 71 | ||
| AK157765.1 | - | 16 - 17 | 1956 | 71 | ||
| BC020013.1 | - | 16 - 17 | 1934 | 3663482 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Noc2l
Genomic Map