Assay Details
| Gene Symbol: Noc2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021303.2 | NP_067278.2 | 8 - 9 | 928 | 78 | |
| GenBank mRNA | AK076287.1 | - | 7 - 8 | 970 | 78 | |
| AK078560.1 | - | 8 - 9 | 904 | 78 | ||
| AK133834.1 | - | 8 - 9 | 928 | 78 | ||
| AK157765.1 | - | 8 - 9 | 921 | 78 | ||
| AK164071.1 | - | 2 - 3 | 2545 | 78 | ||
| AK184541.1 | - | 4 - 5 | 373 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Noc2l
Genomic Map