Assay Details
| Gene Symbol: Stard6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029019.4 | NP_083295.1 | 1 - 2 | 112 | 112 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_029019.4 transcripts.
Set Membership: |
|

Stard6
Genomic Map