Assay Details
| Gene Symbol: Zyg11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033634.3 | NP_001028806.2 | 8 - 9 | 1654 | 82 | |
| GenBank mRNA | AK122545.1 | - | 1 - 2 | 43 | 82 | |
| AK145130.1 | - | 8 - 9 | 1628 | 82 | ||
| AK148335.1 | - | 8 - 9 | 1653 | 82 | ||
| AK148760.1 | - | 2 - 3 | 124 | 82 | ||
| AK166031.1 | - | 8 - 9 | 1634 | 82 | ||
| AK166174.1 | - | 8 - 9 | 1634 | 82 | ||
| AK166278.1 | - | 8 - 9 | 1645 | 82 | ||
| BC139383.1 | - | 8 - 9 | 1621 | 8862405 | 82 | |
| BC139384.1 | - | 8 - 9 | 1621 | 8862406 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zyg11b
Genomic Map