Assay Details
| Gene Symbol: Cdcp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172873.3 | NP_766461.1 | 1 - 2 | 901 | 79 | |
| GenBank mRNA | AK050720.1 | - | 1 - 2 | 902 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdcp2
Genomic Map