Assay Details
| Gene Symbol: Cdcp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172873.3 | NP_766461.1 | 3 - 4 | 1577 | 97 | |
| GenBank mRNA | AK050720.1 | - | 3 - 4 | 1578 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2033116 | AK050720.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2033116 | NM_172873.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 270171 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 294526 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 616489 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 126579 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 603585 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 385946 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 314794 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 13826 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P134561 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P104662 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2033116 | AK050720.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2033116 | NM_172873.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188545 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188561 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188546 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188569 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188534 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188564 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188558 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188548 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188554 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188578 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188551 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188556 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188535 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188568 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188579 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188550 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188565 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188582 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188555 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188580 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188576 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188575 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188537 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188571 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188574 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188583 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188539 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188552 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188573 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188529 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188531 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188544 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23188542 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P104662 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P134561 | - | - |
| Whole Mouse Genome G4122A | A_51_P421551 | AK050720.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P421551 | NM_172873.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119690 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114374 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P104662 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P134561 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04093639 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04027077 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03397243 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04093640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23188546 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188644 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188642 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188590 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03683517 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03423717 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188579 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03243655 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04039067 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03223432 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03821373 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188578 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03837849 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188632 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03990902 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188510 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188659 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188561 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03607389 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188559 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188649 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03393644 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188664 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03607378 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188576 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03607387 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03683515 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03243649 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03201877 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04093640 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03201874 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188658 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188672 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03607383 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03243658 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188544 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188586 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03201870 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03243671 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03607371 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03607386 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188515 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188667 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188534 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188587 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188539 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188628 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188592 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188604 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188678 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03734811 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03734818 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03423718 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03821374 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188662 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188518 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188640 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03223419 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03393645 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04027077 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04027078 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03720015 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03243659 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188643 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188580 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03607396 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188641 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04045580 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188665 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03620973 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188554 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188606 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188660 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188624 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03607373 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03223438 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188565 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03607391 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188511 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188527 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188645 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03734809 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03308137 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188599 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188526 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23188574 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03201875 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03223428 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04093639 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10506641 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10506639 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10506640 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10506642 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10506638 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400007742.mm.1 | NM_172873.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400007742.mm.1 | NM_172873.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400007742.mm.1 | TC0400001121.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400007741.mm.1 | NM_172873.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400007741.mm.1 | NM_172873.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400007741.mm.1 | TC0400001121.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 360365 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2260438 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2260438 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002260438 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1251133 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1251133 | - | - |
| mouse-6 v1.1 (Illumina) | scl25166.4.1_82-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2260438 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl25166.4.1_82-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2260438 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1251133 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1251133 | - | - |

Cdcp2
Genomic Map