Assay Details
| Gene Symbol: Glis1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147221.2 | NP_671754.2 | 3 - 4 | 643 | 132 | |
| GenBank mRNA | JQ043365.1 | - | 2 - 3 | 422 | 132 | |
More Information
Set Membership: |
|

Glis1
Genomic Map