Assay Details
| Gene Symbol: Slc1a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146255.2 | NP_666367.2 | 7 - 8 | 1271 | 65 | |
| GenBank mRNA | AK044586.1 | - | 7 - 8 | 1163 | 65 | |
| AK044671.1 | - | 7 - 8 | 1201 | 65 | ||
| BC030400.1 | - | 7 - 8 | 1282 | 5363389 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc1a7
Genomic Map