Assay Details
| Gene Symbol: Atg9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003917.4 | NP_001003917.2 | 11 - 12 | 2121 | 63 | |
| NM_001288612.1 | NP_001275541.1 | 11 - 12 | 2124 | 63 | ||
| NM_001288613.1 | NP_001275542.1 | 11 - 12 | 2369 | 63 | ||
| NR_109938.1 | 11 - 12 | 2065 | 63 | |||
| GenBank mRNA | AM085510.1 | - | 11 - 12 | 2034 | 63 | |
| BC034139.1 | - | 4 - 5 | 810 | 63 | ||
| BC056488.1 | - | 11 - 12 | 2282 | 63 | ||
| BC079884.1 | - | 11 - 12 | 2010 | 6840050 | 63 | |
| BC082307.1 | - | 9 - 10 | 1826 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atg9a
Genomic Map