Assay Details
| Gene Symbol: Atg9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003917.4 | NP_001003917.2 | 14 - 15 | 2642 | 95 | |
| NM_001288612.1 | NP_001275541.1 | 14 - 15 | 2645 | 95 | ||
| NM_001288613.1 | NP_001275542.1 | 14 - 15 | 2890 | 95 | ||
| NR_109938.1 | 14 - 15 | 2586 | 95 | |||
| GenBank mRNA | AM085510.1 | - | 14 - 15 | 2555 | 95 | |
| BC034139.1 | - | 7 - 8 | 1331 | 95 | ||
| BC056488.1 | - | 14 - 15 | 2803 | 95 | ||
| BC079884.1 | - | 14 - 15 | 2531 | 6840050 | 95 | |
| BC082307.1 | - | 12 - 13 | 2347 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
autophagosome assembly
mitophagy
transport
autophagy
protein transport
negative regulation of interferon-beta production
protein localization to Golgi apparatus
protein localization to pre-autophagosomal structure
piecemeal microautophagy of nucleus
late nucleophagy
innate immune response
molecular_function
autophagosome assembly
mitophagy
transport
autophagy
protein transport
negative regulation of interferon-beta production
protein localization to Golgi apparatus
protein localization to pre-autophagosomal structure
piecemeal microautophagy of nucleus
late nucleophagy
innate immune response
molecular_function
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2424459 | BC079884.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2424459 | BC082307.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2424459 | BC056488.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2424459 | AM085510.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2424459 | AK043624.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 782090 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 35303 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 258102 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 219977 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 10375 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 157413 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 428629 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 29827 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P162952 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2424459 | BC079884.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2424459 | BC082307.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2424459 | BC056488.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2424459 | AM085510.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2424459 | AK043624.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352477 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352484 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352485 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352489 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352481 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352537 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352494 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352535 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352521 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352509 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352520 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352528 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352508 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352523 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352525 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352541 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352488 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352534 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352496 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352515 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352492 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352497 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352540 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352495 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352514 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352510 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352529 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352518 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352479 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352547 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352532 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352544 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352499 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04149747 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P162952 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04149734 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P162952 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00809691 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00114983 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00809671 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00114986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352525 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00114969 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00848357 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00809696 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00114985 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00230145 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352523 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00114984 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00230147 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00526286 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00230151 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352488 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352514 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01142208 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00848365 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00114965 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00809691 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00809688 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00809685 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352524 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352520 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00316215 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352489 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00848370 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00230150 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00316226 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00316221 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00606431 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352503 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352534 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352493 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00316222 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00388320 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00316224 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352543 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00114986 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352528 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00606434 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00606424 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352479 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352532 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352525 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00809671 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00316216 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352499 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10355769 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10355783 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10355764 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10355774 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10355767 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10355780 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10355770 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10355776 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10355765 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10355766 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10355773 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10355768 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10355771 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10355778 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10355781 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10355779 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10355782 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10355772 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10355777 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10355775 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435784_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10355763 | - | - |
| Mouse Genome 430 2.0 Array | 1435784_at | - | - |
| Mouse Genome 430 2.0 Array | 1435784_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 245860_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 245860_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100021129.mm.1 | NM_001003917.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100021129.mm.1 | NM_001288612.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100021129.mm.1 | NM_001288613.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100021129.mm.1 | NR_109938.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100021129.mm.1 | TC0100002602.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100021127.mm.1 | NM_001003917.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100021127.mm.1 | NM_001288612.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100021127.mm.1 | NM_001288613.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100021127.mm.1 | NR_109938.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100021127.mm.1 | TC0100002602.mm.1 | MTA TC |
| Affymetrix HT Mouse Genome 430B Array | 1435784_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435784_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 462333 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 100540286 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 100540286 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2699338 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2699338 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2698801 | - | - |
| mouse-6 v1.1 (Illumina) | scl0245860.3_111-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 100540286 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100540286 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2698801 | - | - |

Atg9a
Genomic Map