Assay Details
| Gene Symbol: Atg9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003917.4 | NP_001003917.2 | 6 - 7 | 643 | 95 | |
| NM_001288612.1 | NP_001275541.1 | 6 - 7 | 646 | 95 | ||
| NM_001288613.1 | NP_001275542.1 | 6 - 7 | 643 | 95 | ||
| NR_109938.1 | 6 - 7 | 646 | 95 | |||
| GenBank mRNA | AK043624.1 | - | 6 - 7 | 697 | 95 | |
| AM085510.1 | - | 6 - 7 | 556 | 95 | ||
| BC056488.1 | - | 6 - 7 | 556 | 95 | ||
| BC079884.1 | - | 6 - 7 | 591 | 6840050 | 95 | |
| BC082307.1 | - | 4 - 5 | 349 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001288613.1 transcripts.
Set Membership: |
|

Atg9a
Genomic Map