Assay Details
| Gene Symbol: Atg9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003917.4 | NP_001003917.2 | 8 - 9 | 1532 | 111 | |
| NM_001288612.1 | NP_001275541.1 | 8 - 9 | 1535 | 111 | ||
| NM_001288613.1 | NP_001275542.1 | 8 - 9 | 1780 | 111 | ||
| NR_109938.1 | 8 - 9 | 1535 | 111 | |||
| GenBank mRNA | AM085510.1 | - | 8 - 9 | 1445 | 111 | |
| BC034139.1 | - | 1 - 2 | 221 | 111 | ||
| BC056488.1 | - | 8 - 9 | 1693 | 111 | ||
| BC079884.1 | - | 8 - 9 | 1480 | 6840050 | 111 | |
| BC082307.1 | - | 6 - 7 | 1238 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atg9a
Genomic Map