Assay Details
| Gene Symbol: Sox5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113559.2 | NP_001107031.2 | 4 - 5 | 764 | 69 | |
| NM_001243163.1 | NP_001230092.1 | 4 - 5 | 659 | 69 | ||
| NM_011444.3 | NP_035574.2 | 4 - 5 | 764 | 69 | ||
| GenBank mRNA | AB006330.1 | - | 4 - 5 | 676 | 69 | |
| AJ010604.1 | - | 4 - 5 | 649 | 69 | ||
| AK029047.1 | - | 7 - 8 | 909 | 69 | ||
| AK081802.1 | - | 4 - 5 | 1056 | 69 | ||
| AK132307.1 | - | 4 - 5 | 657 | 69 | ||
| BC110478.2 | - | 4 - 5 | 648 | 40047865 | 69 | |
| KF793916.1 | - | 4 - 5 | 465 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sox5
Genomic Map