Assay Details
| Gene Symbol: Gpx7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024198.3 | NP_077160.1 | 2 - 3 | 431 | 56 | |
| GenBank mRNA | AK014199.1 | - | 2 - 3 | 429 | 56 | |
| AK084382.1 | - | 2 - 3 | 434 | 56 | ||
| AK165221.1 | - | 2 - 3 | 432 | 56 | ||
| BC003228.1 | - | 2 - 3 | 432 | 3483448 | 56 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23195897 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23195898 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23195893 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23195895 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23195894 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2012960 | AK014199.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2012960 | AK165221.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2012960 | NM_024198.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894534 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 527592 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 436047 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 604503 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 763400 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 421643 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 735124 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 900291 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 677940 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 110210 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 111915 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 18889 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 145249 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 144715 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P122653 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2012960 | AK014199.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2012960 | AK165221.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2012960 | NM_024198.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P461364 | BC003228.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P461364 | AK014199.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P461364 | AK084382.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P461364 | AK165221.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P461364 | NM_024198.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23195902 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23195918 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23195887 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23195919 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23195898 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23195892 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23195888 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23195893 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23195911 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23195903 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23195914 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23195901 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23195886 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23195910 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23195890 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23195899 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23195908 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23195900 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23195896 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23195906 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23195917 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23195909 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23195904 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23195912 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23195895 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23195915 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05353733 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P122653 | - | - |
| Whole Mouse Genome G4122A | A_51_P461364 | BC003228.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P461364 | AK014199.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P461364 | AK084382.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P461364 | AK165221.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P461364 | NM_024198.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107178 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P122653 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05353733 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03748250 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03760653 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03748248 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03748251 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03760661 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03822497 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03760652 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23195897 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23195898 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23195895 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23195893 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23195894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23195871 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03760652 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03760658 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23195904 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23195859 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23195906 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23195876 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03820958 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23195881 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23195888 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23195867 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23195872 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23195832 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23195871 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03896041 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23195889 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23195882 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23195833 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03990523 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04003810 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03995239 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23195840 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03687122 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03652468 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23195873 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03995234 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03427801 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23195900 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23195869 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23195896 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03748251 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23195915 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03427804 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23195850 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23195862 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23195879 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23195914 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23195912 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23195917 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03526334 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23195835 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03300025 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03896044 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03760662 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03820956 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23195886 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23195908 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03888652 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03822497 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23195831 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23195841 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03760661 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23195902 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03672965 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03888650 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10515008 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515011 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515010 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515009 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417836_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10515007 | - | - |
| Mouse Genome 430 2.0 Array | 1417836_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417836_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67305_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67305_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400025817.mm.1 | NM_024198.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400025817.mm.1 | NM_024198.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400025817.mm.1 | TC0400003241.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400025814.mm.1 | NM_024198.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400025814.mm.1 | NM_024198.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400025814.mm.1 | TC0400003241.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400025816.mm.1 | NM_024198.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400025816.mm.1 | NM_024198.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400025816.mm.1 | TC0400003241.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417836_PM_at | - | - |
| Mouse Expression Set 430 | 1417836_at | - | - |
| Murine Genome U74v2 Set | 103607_PM_at | - | - |
| Murine Genome U74v2 Set | 103607_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417836_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417836_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417836_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417836_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 926815 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3440253 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3440253 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003440253 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2724294 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2724294 | - | - |
| mouse-6 v1.1 (Illumina) | scl24011.4.1_9-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3440253 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl24011.4.1_9-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3440253 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2724294 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2724294 | - | - |

Gpx7
Genomic Map