Assay Details
| Gene Symbol: Recql | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204906.1 | NP_001191835.1 | 11 - 12 | 1457 | 79 | |
| NM_001204907.1 | NP_001191836.1 | 11 - 12 | 1457 | 79 | ||
| NM_023042.3 | NP_075529.2 | 11 - 12 | 1457 | 79 | ||
| GenBank mRNA | AB017104.1 | - | 10 - 11 | 1418 | 79 | |
| AB017105.1 | - | 10 - 11 | 1410 | 79 | ||
| AK046660.1 | - | 1 - 2 | 127 | 79 | ||
| AK137589.1 | - | 11 - 12 | 1457 | 79 | ||
| AK138325.1 | - | 11 - 12 | 1413 | 79 | ||
| AK148430.1 | - | 11 - 12 | 1445 | 79 | ||
| AK164344.1 | - | 11 - 12 | 1427 | 79 | ||
| BC014735.1 | - | 10 - 11 | 1389 | 4459244 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Recql
Genomic Map