Assay Details
| Gene Symbol: Recql | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204906.1 | NP_001191835.1 | 13 - 14 | 1760 | 79 | |
| NM_001204907.1 | NP_001191836.1 | 13 - 14 | 1760 | 79 | ||
| NM_023042.3 | NP_075529.2 | 13 - 14 | 1760 | 79 | ||
| GenBank mRNA | AK046660.1 | - | 3 - 4 | 430 | 79 | |
| AK137589.1 | - | 13 - 14 | 1760 | 79 | ||
| AK138325.1 | - | 13 - 14 | 1716 | 79 | ||
| AK138721.1 | - | 13 - 14 | 1801 | 79 | ||
| AK164344.1 | - | 13 - 14 | 1730 | 79 | ||
More Information
Set Membership: |
|

Recql
Genomic Map