Assay Details
| Gene Symbol: Ankzf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026187.5 | NP_080463.3 | 4 - 5 | 921 | 106 | |
| GenBank mRNA | AK156255.1 | - | 4 - 5 | 885 | 106 | |
| AK157405.1 | - | 4 - 5 | 885 | 106 | ||
| BC150879.1 | - | 4 - 5 | 871 | 9087790 | 106 | |
| BU054541.1 | - | 3 - 4 | 322 | 106 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK156255.1 transcripts.
Set Membership: |
|

Ankzf1
Genomic Map