Assay Details
| Gene Symbol: Gm5095 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033454.1 | 4 - 5 | 1380 | 105 | ||
| GenBank mRNA | AK043399.1 | - | 4 - 5 | 1381 | 105 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 9130209A04 gene | 9130209A04Rik | -245.2kb | Sense | Chr. 18 - 47452668 - 47470868 on NCBI Build GRCm38 | |
| Downstream | predicted gene 4146 | Gm4146 | 42.6kb | Anti-sense | Chr. 18 - 47761088 - 47789533 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm5095
Genomic Map