Assay Details
| Gene Symbol: Gm31087 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC049679.1 | - | 209 | 71 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | potassium channel tetramerisation domain containing 16 | Kctd16 |
4930434H12Rik, Gm1267 |
Anti-sense | Chr. 18 - 40258361 - 40531184 on NCBI Build GRCm38 | |
| Downstream | PRELI domain containing 2 | Prelid2 |
1700003A01Rik, C330008K14Rik |
1.6mb | Sense | Chr. 18 - 41875696 - 41951194 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm31087
Genomic Map