Assay Details
| Gene Symbol: 3110002H16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029623.2 | NP_083899.1 | 6 - 7 | 623 | 63 | |
| GenBank mRNA | AK010309.1 | - | 6 - 7 | 617 | 63 | |
| AK013979.1 | - | 1 - 2 | 49 | 63 | ||
| AK144623.1 | - | 6 - 7 | 624 | 63 | ||
| AK154240.1 | - | 6 - 7 | 618 | 63 | ||
| AK159691.1 | - | 6 - 7 | 618 | 63 | ||
| BC012285.1 | - | 6 - 7 | 601 | 63 | ||
| BC021846.1 | - | 6 - 7 | 606 | 5027035 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

3110002H16Rik
Genomic Map