Assay Details
| Gene Symbol: Impact | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008378.2 | NP_032404.1 | 1 - 2 | 113 | 66 | |
| GenBank mRNA | AK138136.1 | - | 1 - 2 | 84 | 66 | |
| BC020524.1 | - | 1 - 2 | 89 | 4223620 | 66 | |
| D87973.1 | - | 1 - 2 | 113 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK138136.1, BC020524.1, D87973.1, NM_008378.2 transcripts.
Set Membership: |
|

Impact
Genomic Map