Assay Details
| Gene Symbol: Gucy2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081076.2 | NP_001074545.1 | 14 - 15 | 2317 | 117 | |
| GenBank mRNA | AK049940.1 | - | 2 - 3 | 336 | 117 | |
| AY395631.1 | - | 14 - 15 | 2317 | 117 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gucy2g
Genomic Map