Assay Details
| Gene Symbol: Gucy2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081076.2 | NP_001074545.1 | 15 - 16 | 2499 | 86 | |
| GenBank mRNA | AK049940.1 | - | 3 - 4 | 518 | 86 | |
| AY395631.1 | - | 15 - 16 | 2499 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gucy2g
Genomic Map