Assay Details
| Gene Symbol: Gucy2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081076.2 | NP_001074545.1 | 2 - 3 | 728 | 62 | |
| GenBank mRNA | AY395631.1 | - | 2 - 3 | 728 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gucy2g
Genomic Map