Assay Details
| Gene Symbol: Dnajc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164583.1 | NP_001158055.1 | 14 - 15 | 2175 | 68 | |
| NM_001164584.1 | NP_001158056.1 | 14 - 15 | 2120 | 68 | ||
| NM_001164585.1 | NP_001158057.1 | 15 - 16 | 2185 | 68 | ||
| NM_198412.2 | NP_940804.1 | 15 - 16 | 2094 | 68 | ||
| GenBank mRNA | AK012831.1 | - | 5 - 6 | 840 | 68 | |
| AK049935.1 | - | 2 - 3 | 137 | 68 | ||
| AK122293.1 | - | 14 - 15 | 2096 | 68 | ||
| AK147570.1 | - | 15 - 16 | 2057 | 68 | ||
| AK147637.1 | - | 15 - 16 | 2057 | 68 | ||
| AK163657.1 | - | 14 - 15 | 1994 | 68 | ||
| BC035262.1 | - | 9 - 10 | 1329 | 68 | ||
| BC060734.1 | - | 14 - 15 | 2175 | 6409942 | 68 | |
| BC064460.1 | - | 15 - 16 | 2063 | 6821822 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dnajc6
Genomic Map