Assay Details
| Gene Symbol: Acp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019800.4 | NP_062774.2 | 3 - 4 | 845 | 71 | |
| GenBank mRNA | AB030038.1 | - | 3 - 4 | 838 | 71 | |
| AK002783.1 | - | 3 - 4 | 759 | 71 | ||
| AK077729.1 | - | 3 - 4 | 833 | 71 | ||
| AK160271.1 | - | 3 - 4 | 764 | 71 | ||
| AK165356.1 | - | 3 - 4 | 846 | 71 | ||
| AK171757.1 | - | 3 - 4 | 760 | 71 | ||
| BC047276.1 | - | 3 - 4 | 585 | 6487551 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acp6
Genomic Map