Assay Details
| Gene Symbol: Polr3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028925.1 | NP_083201.1 | 13 - 14 | 1479 | 61 | |
| GenBank mRNA | AK016716.1 | - | 13 - 14 | 1506 | 61 | |
| BC147485.1 | - | 13 - 14 | 1479 | 9056238 | 61 | |
| BC151028.1 | - | 13 - 14 | 1479 | 9087941 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Molecular Function -
Gene Ontology Categories:
Function(s)
Process(es)
immune system process
transcription, DNA-templated
positive regulation of interferon-beta production
innate immune response
positive regulation of innate immune response
defense response to virus
RNA polymerase III activity
DNA binding
DNA-directed RNA polymerase activity
transferase activity
nucleotidyltransferase activity
immune system process
transcription, DNA-templated
positive regulation of interferon-beta production
innate immune response
positive regulation of innate immune response
defense response to virus
RNA polymerase III activity
DNA binding
DNA-directed RNA polymerase activity
transferase activity
nucleotidyltransferase activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054739 | BC147485.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054739 | BC151028.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054739 | NM_028925.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054739 | AK016716.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931641 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 952709 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 145956 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 895605 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 519534 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 169405 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 613820 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894974 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 148846 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 367037 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 49263 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 58701 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P112751 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P138976 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054739 | BC147485.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054739 | BC151028.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054739 | NM_028925.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054739 | AK016716.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P165156 | AK016716.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P309008 | AK212331.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P309008 | BC026793.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P309008 | BC151028.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P309008 | NM_028925.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P309008 | AK016716.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04998454 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P138976 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P112751 | - | - |
| Whole Mouse Genome G4122A | A_52_P309008 | AK212331.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P309008 | BC026793.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P309008 | BC151028.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P309008 | NM_028925.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P309008 | AK016716.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P165156 | AK016716.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P112751 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P138976 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22350815 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22350812 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22350818 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22350817 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02349880 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10500389 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500391 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500397 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500394 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500396 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500403 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500401 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500392 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500390 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500395 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500393 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500402 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500398 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500399 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500400 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451658_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453256_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456803_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10500388 | - | - |
| Mouse Genome 430 2.0 Array | 1453256_at | - | - |
| Mouse Genome 430 2.0 Array | 1451658_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1451658_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1453256_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1456803_at | - | - |
| Mouse Genome 430 2.0 Array | 1456803_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74414_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74414_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300019678.mm.1 | NM_028925.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019678.mm.1 | NM_028925.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019678.mm.1 | TC0300002527.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300019677.mm.1 | NM_028925.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019677.mm.1 | NM_028925.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019677.mm.1 | TC0300002527.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1453256_at | - | - |
| Mouse Expression Set 430 | 1451658_a_at | - | - |
| Mouse Expression Set 430 | 1451658_PM_a_at | - | - |
| Mouse Expression Set 430 | 1453256_PM_at | - | - |
| Mouse Expression Set 430 | 1456803_at | - | - |
| Mouse Expression Set 430 | 1456803_PM_at | - | - |
| Murine Genome U74v2 Set | 131197_PM_at | - | - |
| Murine Genome U74v2 Set | 131197_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451658_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1453256_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451658_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1453256_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451658_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451658_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1453256_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1453256_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456803_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456803_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 104200050 | - | - |
| MouseWG-6 v2 Expression BeadChip | 6940546 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 104200050 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2696471 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1231253 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2696471 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 104200050 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104200050 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1231253 | - | - |

Polr3c
Genomic Map