Assay Details
| Gene Symbol: Polr3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028925.1 | NP_083201.1 | 4 - 5 | 692 | 108 | |
| GenBank mRNA | AK016716.1 | - | 4 - 5 | 719 | 108 | |
| AK043194.1 | - | 3 - 4 | 827 | 108 | ||
| AK212331.1 | - | 1 - 2 | 55 | 108 | ||
| BC147485.1 | - | 4 - 5 | 9056238 | 108 | ||
| BC151028.1 | - | 4 - 5 | 692 | 9087941 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Polr3c
Genomic Map