Assay Details
| Gene Symbol: Gpr19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167694.2 | NP_001161166.1 | 3 - 4 | 300 | 85 | |
| NM_001167699.2 | NP_001161171.1 | 1 - 2 | 230 | 85 | ||
| GenBank mRNA | BC021648.1 | - | 3 - 4 | 239 | 3989077 | 85 |
| DV660907.1 | - | 3 - 4 | 234 | 85 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC021648.1, NM_001167694.2, NM_001167699.2 transcripts.
Set Membership: |
|

Gpr19
Genomic Map