Assay Details
| Gene Symbol: Adnp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175028.1 | NP_778193.1 | 2 - 3 | 231 | 77 | |
| GenBank mRNA | AB093268.1 | - | 2 - 3 | 231 | 77 | |
| BC044898.1 | - | 2 - 3 | 136 | 77 | ||
| BC047352.1 | - | 2 - 3 | 136 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adnp2
Genomic Map