Assay Details
| Gene Symbol: Sstr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009216.3 | NP_033242.1 | 1 - 2 | 79 | 112 | |
| GenBank mRNA | AK046464.1 | - | 1 - 2 | 79 | 112 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK046464.1, NM_009216.3 transcripts.
Set Membership: |
|

Sstr1
Genomic Map