| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100015756.mm.1 |
NM_001160711.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015756.mm.1 |
NM_001160711.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015756.mm.1 |
NM_001160712.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015756.mm.1 |
NM_001160712.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015756.mm.1 |
NM_001160713.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015756.mm.1 |
NM_001160713.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015756.mm.1 |
NM_027987.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015756.mm.1 |
NM_027987.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015756.mm.1 |
TC1100001677.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100015754.mm.1 |
NM_001160711.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015754.mm.1 |
NM_001160711.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015754.mm.1 |
NM_001160712.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015754.mm.1 |
NM_001160712.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015754.mm.1 |
NM_001160713.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015754.mm.1 |
NM_001160713.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015754.mm.1 |
NM_027987.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015754.mm.1 |
NM_027987.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015754.mm.1 |
TC1100001677.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100015755.mm.1 |
NM_001160711.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015755.mm.1 |
NM_001160711.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015755.mm.1 |
NM_001160712.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015755.mm.1 |
NM_001160712.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015755.mm.1 |
NM_001160713.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015755.mm.1 |
NM_001160713.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015755.mm.1 |
NM_027987.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015755.mm.1 |
NM_027987.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015755.mm.1 |
TC1100001677.mm.1 |
MTA TC |