Assay Details
| Gene Symbol: Cd300lg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK171982.1 | - | 2 - 3 | 404 | 88 | |
| BC119315.1 | - | 2 - 3 | 407 | 8734064 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cd300lg
Genomic Map