Assay Details
| Gene Symbol: Cd300lg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160711.1 | NP_001154183.1 | 6 - 7 | 1087 | 79 | |
| NM_001160712.1 | NP_001154184.1 | 6 - 7 | 835 | 79 | ||
| NM_001160713.1 | NP_001154185.1 | 5 - 6 | 955 | 79 | ||
| NM_027987.3 | NP_082263.2 | 5 - 6 | 703 | 79 | ||
| GenBank mRNA | AB243063.1 | - | 6 - 7 | 1050 | 79 | |
| AB243064.1 | - | 5 - 6 | 918 | 79 | ||
| AB243065.1 | - | 6 - 7 | 798 | 79 | ||
| AB243066.1 | - | 5 - 6 | 666 | 79 | ||
| AK009375.1 | - | 5 - 6 | 666 | 79 | ||
| AK171982.1 | - | 5 - 6 | 647 | 79 | ||
| AY457055.1 | - | 5 - 6 | 623 | 79 | ||
| BC116906.1 | - | 5 - 6 | 650 | 40126225 | 79 | |
| BC119315.1 | - | 5 - 6 | 650 | 8734064 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cd300lg
Genomic Map