Assay Details
| Gene Symbol: Eif3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010123.3 | NP_034253.3 | 12 - 13 | 2151 | 86 | |
| GenBank mRNA | AK013070.1 | - | 2 - 3 | 466 | 86 | |
| AK145132.1 | - | 12 - 13 | 2159 | 86 | ||
| AK145629.1 | - | 12 - 13 | 2116 | 86 | ||
| AK149482.1 | - | 12 - 13 | 2104 | 86 | ||
| AK162784.1 | - | 1 - 2 | 64 | 86 | ||
| AK167010.1 | - | 12 - 13 | 2118 | 86 | ||
| X17373.1 | - | 5 - 6 | 771 | 86 | ||
| X84651.1 | - | 5 - 6 | 771 | 86 | ||
More Information
Set Membership: |
|

Eif3a
Genomic Map