Assay Details
| Gene Symbol: Prdx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007452.2 | NP_031478.1 | 2 - 3 | 245 | 104 | |
| GenBank mRNA | AK002448.1 | - | 2 - 3 | 179 | 104 | |
| BC005626.1 | - | 2 - 3 | 196 | 3963113 | 104 | |
| BY318475.1 | - | 2 - 3 | 248 | 104 | ||
| M28723.1 | - | 2 - 3 | 221 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prdx3
Genomic Map