Assay Details
| Gene Symbol: Gnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160017.1 | NP_001153489.1 | 1 - 2 | 220 | 90 | |
| GenBank mRNA | AK156101.1 | - | 1 - 2 | 217 | 90 | |
| BG807492.1 | - | 1 - 2 | 211 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK156101.1, NM_001160017.1 transcripts.
Set Membership: |
|

Gnb1
Genomic Map