Assay Details
Gene Symbol: Tnfrsf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011659.2 | NP_035789.1 | 2 - 3 | 429 | 136 | |
GenBank mRNA | BC065782.1 | - | 2 - 3 | 268 | 136 | |
BC139239.1 | - | 2 - 3 | 299 | 8862261 | 136 | |
BC139266.1 | - | 2 - 3 | 299 | 8862288 | 136 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
