| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400034802.mm.1 |
NM_001256107.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034802.mm.1 |
NM_001256107.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034802.mm.1 |
NM_001256108.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034802.mm.1 |
NM_145124.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034802.mm.1 |
NM_145124.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034802.mm.1 |
TC0400004147.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400034801.mm.1 |
NM_001256107.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034801.mm.1 |
NM_001256107.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034801.mm.1 |
NM_001256108.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034801.mm.1 |
NM_145124.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034801.mm.1 |
NM_145124.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034801.mm.1 |
TC0400004147.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400034805.mm.1 |
NM_001256107.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034805.mm.1 |
NM_001256107.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034805.mm.1 |
NM_001256108.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034805.mm.1 |
NM_145124.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034805.mm.1 |
NM_145124.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034805.mm.1 |
TC0400004147.mm.1 |
MTA TC |