| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700025698.mm.1 |
NM_011274.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700025698.mm.1 |
NM_011274.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700025698.mm.1 |
TC0700002873.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700025697.mm.1 |
NM_011274.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700025697.mm.1 |
NM_011274.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700025697.mm.1 |
TC0700002873.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700025700.mm.1 |
NM_011274.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700025700.mm.1 |
NM_011274.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700025700.mm.1 |
TC0700002873.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700025699.mm.1 |
NM_011274.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700025699.mm.1 |
NM_011274.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700025699.mm.1 |
TC0700002873.mm.1 |
MTA TC |