Assay Details
| Gene Symbol: Trim36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170855.1 | NP_001164326.1 | 4 - 5 | 1017 | 88 | |
| NM_178872.4 | NP_849203.2 | 4 - 5 | 1018 | 88 | ||
| GenBank mRNA | AB103063.1 | - | 4 - 5 | 812 | 88 | |
| AY251389.1 | - | 4 - 5 | 775 | 88 | ||
| BC058106.1 | - | 4 - 5 | 936 | 6836197 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trim36
Genomic Map