Assay Details
| Gene Symbol: Trim36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170855.1 | NP_001164326.1 | 6 - 7 | 1355 | 85 | |
| NM_178872.4 | NP_849203.2 | 6 - 7 | 1356 | 85 | ||
| GenBank mRNA | AY251389.1 | - | 6 - 7 | 1113 | 85 | |
| BC058106.1 | - | 6 - 7 | 1274 | 6836197 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trim36
Genomic Map