Assay Details
| Gene Symbol: Phf13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172705.2 | NP_766293.2 | 2 - 3 | 456 | 84 | |
| GenBank mRNA | AK132066.1 | - | 2 - 3 | 461 | 84 | |
| AY069976.2 | - | 2 - 3 | 381 | 84 | ||
| BC029632.1 | - | 2 - 3 | 456 | 3500487 | 84 | |
| BC059282.1 | - | 2 - 3 | 456 | 3500487 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Phf13
Genomic Map