Assay Details
| Gene Symbol: Ifrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013562.2 | NP_038590.2 | 8 - 9 | 1129 | 88 | |
| GenBank mRNA | AK078888.1 | - | 1127 | 88 | ||
| AK168682.1 | - | 8 - 9 | 1126 | 87 | ||
| AK219989.1 | - | 4 - 5 | 355 | 88 | ||
| BC037013.1 | - | 8 - 9 | 1103 | 88 | ||
| BC043723.1 | - | 8 - 9 | 1096 | 5369100 | 88 | |
| X17400.1 | - | 8 - 9 | 1106 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ifrd1
Genomic Map