Assay Details
| Gene Symbol: Spink6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013797.1 | NP_001013819.1 | 2 - 3 | 154 | 87 | |
| GenBank mRNA | AK028032.1 | - | 2 - 3 | 154 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Spink6
Genomic Map