Assay Details
| Gene Symbol: Arhgdib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007486.5 | NP_031512.1 | 1 - 2 | 102 | 86 | |
| GenBank mRNA | AK002516.1 | - | 1 - 2 | 64 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK002516.1, NM_007486.5 transcripts.
Set Membership: |
|

Arhgdib
Genomic Map